siRNA Details
Gene Symbol: PAXIP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024476.1 | 3 | 860 | |||
NR_024477.1 | 3 | 860 | ||||
GenBank mRNA | AK056609.1 | - | 3 | 850 | ||
AK311721.1 | - | 2 | 783 | |||
BC033053.1 | - | 3 | 886 | |||
BC050402.1 | - | 3 | 845 |
