siRNA Details
Gene Symbol: FAM92A1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003612.2 | 1 | 2090 | |||
GenBank mRNA | BC022019.1 | - | 1 | 2053 | ||
BC059396.1 | - | 1 | 2090 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ429004.1 | - | 1 | 322 | Chr. 4 - 183039434 - 183040093 [+] on Build GRCh38 |
