siRNA Details
Gene Symbol: TTTY11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001548.2 | 4 | 688 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF332240.3 | - | 4 | 688 | Chr. Y - 8783310 - 8817382 [-] on Build GRCh38 |
