siRNA Details
Gene Symbol: TTTY13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001537.1 | 5 | 529 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF332242.3 | - | 5 | 529 | Chr. Y - 21583600 - 21594666 [-] on Build GRCh38 |
