siRNA Details
Gene Symbol: CTBP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033339.1 | 2 | 932 | |||
GenBank mRNA | AK056133.1 | - | 1 | 1578 | ||
AK125775.1 | - | 3 | 886 | |||
BC013775.1 | - | 1 | 35 | |||
BC111466.1 | - | 1 | 380 | |||
BC120947.1 | - | 1 | 484 | 40115129 | ||
BC120948.1 | - | 1 | 484 | 40115130 |
