siRNA Details
Gene Symbol: PICSAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024089.2 | 2 | 586 | |||
GenBank mRNA | BC027456.1 | - | 2 | 586 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PICSAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024089.2 | 2 | 586 | |||
GenBank mRNA | BC027456.1 | - | 2 | 586 |