siRNA Details
Gene Symbol: HIST2H2BA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027337.1 | 1, 2 | 385 | |||
GenBank mRNA | BC106916.2 | - | 1, 2 | 385 | ||
BC126904.1 | - | 1, 2 | 386 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF887452.1 | - | 1, 2 | 391 | Chr. 1 - 121108210 - 121117257 [-] on Build GRCh38 |
