siRNA Details
Gene Symbol: LINC00894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027456.1 | 8, 9 | 2511 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD558604.1 | - | 1, 2 | 211 | Chr. X - 149960760 - 149963204 [+] on Build GRCh38 |
