siRNA Details
Gene Symbol: LINC00894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027456.1 | 12 | 3107 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB523915.1 | - | 3 | 376 | Chr. X - 150016034 - 150016787 [+] on Build GRCh38 |
