siRNA Details
Gene Symbol: FAM186B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027450.1 | 7 | 2908 | |||
XM_006719625.2 | XP_006719688.1 | 8 | 2931 | |||
XM_006719626.2 | XP_006719689.1 | 7 | 2740 | |||
XM_011538797.2 | XP_011537099.1 | 6 | 2570 | |||
XM_017020008.1 | XP_016875497.1 | 8 | 3182 | |||
GenBank mRNA | AK302702.1 | - | 7 | 2908 |
