siRNA Details
Gene Symbol: LINC00710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015413.1 | 6 | 1007 | |||
NR_024410.1 | 5 | 937 | ||||
NR_024411.1 | 4 | 826 | ||||
GenBank mRNA | BC033338.1 | - | 6 | 939 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB508949.2 | - | 3 | 581 | Chr. 10 - 10934950 - 10946576 [-] on Build GRCh38 |
