siRNA Details
Gene Symbol: ST7-OT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002329.1 | 5 | 1439 | |||
GenBank mRNA | BC043268.2 | - | 5 | 1439 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM413626.1 | - | 5 | 1438 | Chr. 7 - 116953898 - 116959817 [+] on Build GRCh38 |
