siRNA Details
Gene Symbol: SNORA65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002449.2 | 1 | 91 | |||
GenBank mRNA | Y11159.1 | - | 1 | 91 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNORA65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002449.2 | 1 | 91 | |||
GenBank mRNA | Y11159.1 | - | 1 | 91 |