siRNA Details
Gene Symbol: RNU4ATAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023343.1 | 1 | 69 | |||
GenBank mRNA | U62822.1 | - | 1 | 69 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNU4ATAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023343.1 | 1 | 69 | |||
GenBank mRNA | U62822.1 | - | 1 | 69 |