siRNA Details
Gene Symbol: NXF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002216.1 | 3 | 351 | |||
GenBank mRNA | AK124700.1 | - | 3 | 351 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA187251.1 | - | 3 | 351 | Chr. X - 102549965 - 102562686 [+] on Build GRCh38 |
