siRNA Details
Gene Symbol: NXF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002216.1 | 15 | 1762 | |||
GenBank mRNA | AK124700.1 | - | 15 | 1762 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NXF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002216.1 | 15 | 1762 | |||
GenBank mRNA | AK124700.1 | - | 15 | 1762 |