siRNA Details
Gene Symbol: LINC00521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024182.1 | 2 | 200 | |||
NR_024183.1 | 2 | 452 | ||||
NR_024184.1 | 2 | 452 | ||||
GenBank mRNA | AK097741.1 | - | 2 | 447 | ||
BC021728.2 | - | 2 | 451 | |||
BC031252.1 | - | 2 | 200 | |||
BI458986.1 | - | 2 | 456 | |||
DB446670.1 | - | 2 | 80 |
