siRNA Details
Gene Symbol: LINC00521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024182.1 | 8 | 1363 | |||
NR_024183.1 | 7 | 1138 | ||||
NR_024184.1 | 6 | 988 | ||||
GenBank mRNA | AK097741.1 | - | 7 | 1133 | ||
BC021728.2 | - | 6 | 987 | |||
BC031252.1 | - | 8 | 1363 |
