siRNA Details
Gene Symbol: KC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002838.1 | 3 | 364 | |||
GenBank mRNA | AY762618.1 | - | 3 | 364 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002838.1 | 3 | 364 | |||
GenBank mRNA | AY762618.1 | - | 3 | 364 |