siRNA Details
Gene Symbol: MAFG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015454.1 | 2 | 635 | |||
GenBank mRNA | BC038198.1 | - | 2 | 63 | ||
BC046361.1 | - | 2 | 522 | |||
BC101984.1 | - | 2 | 635 | |||
BC101987.1 | - | 2 | 635 | |||
BC104740.1 | - | 2 | 635 | |||
BC134411.1 | - | 2 | 635 |
