siRNA Details
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147200.2 | NP_671733.2 | 2 | 198 | ||
XM_011535386.1 | XP_011533688.1 | 2 | 1133 | |||
GenBank mRNA | AF085921.1 | - | 2 | 106 | ||
AF136405.1 | - | 2 | 106 | |||
AK310922.1 | - | 2 | 129 | |||
AK314415.1 | - | 2 | 121 |
