siRNA Details
Gene Symbol: LINC01348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027454.2 | 1 | 271 | |||
GenBank mRNA | AK128489.1 | - | 2 | 881 | ||
AV652367.1 | - | 1 | 161 | |||
BX100191.1 | - | 1 | 271 | |||
DA457760.1 | - | 1 | 88 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG568342.1 | - | 1 | 249 | Chr. 1 - 235067895 - 235069796 [-] on Build GRCh38 |
