siRNA Details
Gene Symbol: LINC01348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027454.2 | 2 | 523 | |||
GenBank mRNA | AK128489.1 | - | 2 | 2035 | ||
DA457760.1 | - | 2 | 340 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG568342.1 | - | 2 | 461 | Chr. 1 - 235067895 - 235069796 [-] on Build GRCh38 |
