siRNA Details
Gene Symbol: LINC01348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027454.2 | 1, 2 | 424 | |||
GenBank mRNA | AK128489.1 | - | 2 | 1936 | ||
AV652367.1 | - | 1, 2 | 314 | |||
BX100191.1 | - | 1, 2 | 424 | |||
DA457760.1 | - | 1, 2 | 241 |
