siRNA Details
Gene Symbol: MIR7-3HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027148.1 | 3 | 624 | |||
GenBank mRNA | AB058892.2 | - | 3 | 589 | ||
AB058893.2 | - | 5 | 758 | |||
BC090884.1 | - | 3 | 620 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR7-3HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027148.1 | 3 | 624 | |||
GenBank mRNA | AB058892.2 | - | 3 | 589 | ||
AB058893.2 | - | 5 | 758 | |||
BC090884.1 | - | 3 | 620 |