siRNA Details
Gene Symbol: LINC01547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027128.1 | 3 | 705 | |||
NR_027129.1 | 4 | 849 | ||||
GenBank mRNA | AF380178.1 | - | 4 | 781 | ||
AY040088.1 | - | 5 | 731 | |||
BC128244.1 | - | 3 | 563 | |||
BC128245.1 | - | 3 | 563 | |||
BI829356.1 | - | 3 | 669 | |||
BI907880.1 | - | 3 | 705 |
