siRNA Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026541.1 | 2 | 154 | |||
NR_026563.1 | 1 | 299 | ||||
NR_026564.1 | 2 | 154 | ||||
GenBank mRNA | AK131059.1 | - | 2 | 131 | ||
BC026079.1 | - | 1 | 305 | |||
BC026082.1 | - | 2 | 150 | |||
BC034967.1 | - | 1 | 305 | |||
BC044592.1 | - | 1 | 298 | |||
BC056262.1 | - | 2 | 154 | |||
DB462574.1 | - | 2 | 154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY026050.1 | - | 2 | 155 | Chr. 11 - 18209138 - 18209900 [+] on Build GRCh38 |
