siRNA Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026541.1 | 2 | 839 | |||
NR_026563.1 | 1 | 984 | ||||
NR_026564.1 | 2 | 839 | ||||
GenBank mRNA | AK131059.1 | - | 2 | 817 | ||
BC026079.1 | - | 1 | 991 | |||
BC026082.1 | - | 2 | 836 | |||
BC034967.1 | - | 1 | 991 | |||
BC044592.1 | - | 1 | 984 | |||
BC056262.1 | - | 2 | 841 |
