siRNA Details
Gene Symbol: SNHG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027058.1 | 3 | 690 | |||
GenBank mRNA | AF130050.1 | - | 2 | 315 | ||
BX648413.1 | - | 3 | 650 | |||
FN357115.1 | - | 3 | 531 | |||
FN357116.1 | - | 3 | 401 | |||
FN357117.1 | - | 3 | 584 |
