siRNA Details
Gene Symbol: SNHG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027058.1 | 3 | 2015 | |||
GenBank mRNA | AF130050.1 | - | 2 | 1640 | ||
AK098448.1 | - | 1 | 762 | |||
BX648413.1 | - | 3 | 1974 | |||
FN357117.1 | - | 3 | 1909 | |||
FN357118.1 | - | 4 | 1187 | |||
FN357119.1 | - | 3 | 1083 |
