siRNA Details
Gene Symbol: ZNF295-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027273.2 | 2 | 612 | |||
NR_119384.1 | 2 | 841 | ||||
NR_119385.1 | 2 | 658 | ||||
GenBank mRNA | AK055167.1 | - | 1 | 2672 | ||
BC029588.1 | - | 2 | 612 |
