siRNA Details
Gene Symbol: KRTAP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021489.1 | 2 | 2245 | |||
GenBank mRNA | AK055141.1 | - | 2 | 1936 |
Gene Symbol: LOC105375030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096675.1 | - | 1 | 770 |
