siRNA Details
Gene Symbol: WDFY3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015359.1 | 1 | 298 | |||
GenBank mRNA | AY250185.1 | - | 2 | 651 | ||
BC019264.1 | - | 2 | 204 | |||
BC033127.1 | - | 1 | 279 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WDFY3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015359.1 | 1 | 298 | |||
GenBank mRNA | AY250185.1 | - | 2 | 651 | ||
BC019264.1 | - | 2 | 204 | |||
BC033127.1 | - | 1 | 279 |