siRNA Details
Gene Symbol: LINC01134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024455.1 | 2, 3 | 295 | |||
GenBank mRNA | BC016994.1 | - | 2, 3 | 283 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG032857.1 | - | 2, 3 | 282 | Chr. 1 - 3900417 - 3914157 [+] on Build GRCh38 |
