siRNA Details
Gene Symbol: LINC01134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024455.1 | 4 | 1095 | |||
GenBank mRNA | AK130173.1 | - | 2 | 425 | ||
BC016994.1 | - | 4 | 1083 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024455.1 | 4 | 1095 | |||
GenBank mRNA | AK130173.1 | - | 2 | 425 | ||
BC016994.1 | - | 4 | 1083 |