siRNA Details
Gene Symbol: CCDC183-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024580.1 | 5 | 918 | |||
GenBank mRNA | AK055547.1 | - | 5 | 918 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC183-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024580.1 | 5 | 918 | |||
GenBank mRNA | AK055547.1 | - | 5 | 918 |