siRNA Details
Gene Symbol: MIR503HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024607.1 | 3 | 756 | |||
GenBank mRNA | AW378319.1 | - | 1 | 394 | ||
CN297454.1 | - | 3 | 665 | |||
KM886853.1 | - | 3 | 637 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX116517.1 | - | 3 | 674 | Chr. X - 134543340 - 134546618 [-] on Build GRCh38 |
