siRNA Details
Gene Symbol: LINC00839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026827.1 | 4 | 797 | |||
GenBank mRNA | AK090789.1 | - | 4 | 744 | ||
AK092104.1 | - | 4 | 747 | |||
BI517331.1 | - | 4 | 541 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026827.1 | 4 | 797 | |||
GenBank mRNA | AK090789.1 | - | 4 | 744 | ||
AK092104.1 | - | 4 | 747 | |||
BI517331.1 | - | 4 | 541 |