siRNA Details
Gene Symbol: LINC00839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026827.1 | 1 | 14 | |||
GenBank mRNA | AK293138.1 | - | 1 | 14 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DC297724.1 | - | 1 | 14 | Chr. 10 - 42475491 - 42477459 [+] on Build GRCh38 |
