siRNA Details
Gene Symbol: SERPINA13P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015340.1 | 5 | 1684 | |||
GenBank mRNA | AY358238.1 | - | 5 | 1684 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SERPINA13P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015340.1 | 5 | 1684 | |||
GenBank mRNA | AY358238.1 | - | 5 | 1684 |