siRNA Details
Gene Symbol: LINC01550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015430.1 | 3 | 509 | |||
GenBank mRNA | BC043585.1 | - | 3 | 509 | ||
JX073283.1 | - | 3 | 445 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI834723.1 | - | 3 | 512 | Chr. 14 - 97926352 - 97978124 [-] on Build GRCh38 |
