siRNA Details
Gene Symbol: SNHG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003672.2 | 2 | 987 | |||
NR_024542.1 | 3 | 358 | ||||
NR_024543.1 | 3 | 358 | ||||
GenBank mRNA | AK090585.1 | - | 2 | 982 | ||
BC042369.1 | - | 3 | 378 | |||
BC042667.1 | - | 2 | 1001 |
Gene Symbol: SNORA17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054908.1 | - | 3 | 358 |
