siRNA Details
Gene Symbol: LOC650226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029420.1 | 6 | 1236 |
Gene Symbol: LOC105375816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517651.2 | XP_011515953.1 | 2 | 977 |
