siRNA Details
Gene Symbol: URB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026845.1 | 1 | 266 | |||
GenBank mRNA | AK026376.1 | - | 1 | 244 | ||
BC007928.1 | - | 1 | 272 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI756790.1 | - | 1 | 241 | Chr. 21 - 32393158 - 32393962 [+] on Build GRCh38 |
