siRNA Details
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024127.1 | 2, 3 | 376 | |||
GenBank mRNA | AF277181.1 | - | 2, 3 | 164 | ||
BC022205.1 | - | 2, 3 | 1005 | |||
BC064589.1 | - | 2, 3 | 364 | |||
BC068048.1 | - | 2, 3 | 376 | |||
KJ782215.1 | - | 3, 4 | 1505 |
