siRNA Details
Gene Symbol: LINC01102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015399.1 | 4 | 1369 | |||
GenBank mRNA | AL137513.1 | - | 4 | 1337 | ||
BC105699.1 | - | 4 | 1263 | |||
BC105700.1 | - | 4 | 1264 | |||
BC105701.1 | - | 4 | 1263 | |||
BC105702.1 | - | 4 | 1263 |
