siRNA Details
Gene Symbol: LINC01102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015399.1 | 2 | 388 | |||
GenBank mRNA | AK055090.1 | - | 2 | 373 | ||
AK126958.1 | - | 2 | 1468 | |||
AL137513.1 | - | 2 | 357 | |||
BC105699.1 | - | 2 | 282 | |||
BC105700.1 | - | 2 | 282 | |||
BC105701.1 | - | 2 | 282 | |||
BC105702.1 | - | 2 | 282 | |||
DA506317.1 | - | 2 | 389 |
