siRNA Details
Gene Symbol: UBE2Q2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003661.2 | 4 | 1476 | |||
GenBank mRNA | AK125266.1 | - | 4 | 1476 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN414393.1 | - | 5 | 478 | Chr. 15 - 84542144 - 84570712 [-] on Build GRCh38 |
