siRNA Details
Gene Symbol: NKAPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027131.1 | 1 | 114 | |||
GenBank mRNA | AK093895.1 | - | 1 | 80 | ||
BC022359.1 | - | 1 | 54 | |||
BC113730.1 | - | 1 | 53 | |||
BC113732.1 | - | 1 | 53 | |||
BX537594.1 | - | 1 | 106 |
