siRNA Details
Gene Symbol: MAP3K14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024434.2 | 3 | 344 | |||
NR_024435.2 | 4 | 798 | ||||
NR_110324.1 | 3 | 529 | ||||
NR_110325.1 | 5 | 524 | ||||
NR_110326.1 | 4 | 428 | ||||
GenBank mRNA | AK311429.1 | - | 3 | 333 | ||
AK311564.1 | - | 3 | 416 | |||
AL710937.1 | - | 5 | 495 | |||
BC031942.1 | - | 4 | 687 | |||
DB294632.1 | - | 4 | 390 |
